Assay Details
| Gene Symbol: C4H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204390.1 | NP_001191319.1 | 5 - 6 | 761 | 71 | |
| GenBank mRNA | AK231836.1 | - | 4 - 5 | 520 | 71 | |
| AK233670.1 | - | 5 - 6 | 729 | 71 | ||
| AK234116.1 | - | 5 - 6 | 729 | 71 | ||
| AK351176.1 | - | 5 - 6 | 656 | 71 | ||
| AK397935.1 | - | 5 - 6 | 804 | 71 | ||
| AY609895.1 | - | 5 - 6 | 654 | 71 | ||
| EU814887.1 | - | 5 - 6 | 730 | 71 | ||
More Information
Set Membership: |
|

C4H1orf43
Genomic Map