Assay Details
| Gene Symbol: IFIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204395.2 | NP_001191324.1 | 2 | 1558 | 77 | |
| XM_021071929.1 | XP_020927588.1 | 1717 | 77 | |||
| GenBank mRNA | AK343662.1 | - | 2 | 1515 | 77 | |
| HQ200930.1 | - | 1 | 1450 | 77 | ||
More Information
Set Membership: |
|

IFIT3
Genomic Map