Assay Details
| Gene Symbol: BMPR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204767.1 | NP_001191696.1 | 7 - 8 | 874 | 74 | |
| XM_005671180.3 | XP_005671237.1 | 1043 | 74 | |||
| XM_021072031.1 | XP_020927690.1 | 1436 | 74 | |||
| XM_021072032.1 | XP_020927691.1 | 1399 | 74 | |||
| XM_021072033.1 | XP_020927692.1 | 1636 | 74 | |||
| GenBank mRNA | HQ180178.1 | - | 7 - 8 | 874 | 74 | |
More Information
Set Membership: |
|

BMPR1A
Genomic Map