Assay Details
| Gene Symbol: SLA-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097427.1 | NP_001090896.1 | 8 | 1457 | 87 | |
| XM_021097849.1 | XP_020953508.1 | 1567 | 87 | |||
| XM_021097850.1 | XP_020953509.1 | 1449 | 87 | |||
| GenBank mRNA | AF464009.1 | - | 8 | 1443 | 87 | |
| AF464010.1 | - | 8 | 1457 | 87 | ||
| AF464040.1 | - | 8 | 1406 | 87 | ||
| AK232238.1 | - | 8 | 1438 | 87 | ||
| AK236896.1 | - | 8 | 1429 | 87 | ||
| AK392851.1 | - | 8 | 1458 | 87 | ||
| AK394395.1 | - | 8 | 1405 | 87 | ||
| AK395948.1 | - | 8 | 1458 | 87 | ||
| AY247769.1 | - | 8 | 1414 | 87 | ||
| AY247770.1 | - | 8 | 1414 | 87 | ||
| DQ303224.1 | - | 8 | 1375 | 87 | ||
| DQ303227.1 | - | 8 | 1375 | 87 | ||
| DQ883212.1 | - | 8 | 1405 | 87 | ||
| EU432091.1 | - | 8 | 1375 | 87 | ||
| EU432093.1 | - | 8 | 1375 | 87 | ||
| FJ883489.1 | - | 8 | 1384 | 87 | ||
| KU754574.1 | - | 8 | 1396 | 87 | ||
| KU754578.1 | - | 8 | 1396 | 87 | ||
| KU754579.1 | - | 8 | 1405 | 87 | ||
| KU754580.1 | - | 8 | 1405 | 87 | ||
More Information
Set Membership: |
|

SLA-3
Genomic Map