Assay Details
| Gene Symbol: RFX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094829.1 | XP_020950488.1 | 755 | 67 | ||
| XM_021094830.1 | XP_020950489.1 | 669 | 67 | |||
| XM_021094836.1 | XP_020950495.1 | 666 | 67 | |||
| XM_021094844.1 | XP_020950503.1 | 1184 | 67 | |||
| XM_021094854.1 | XP_020950513.1 | 1015 | 67 | |||
| XM_021094860.1 | XP_020950519.1 | 3458 | 67 | |||
More Information
Set Membership: |
|

RFX7
Genomic Map