Assay Details
| Gene Symbol: IGFBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005156.1 | NP_001005156.1 | 1 - 2 | 519 | 119 | |
| GenBank mRNA | AF085482.1 | - | 1 - 2 | 519 | 119 | |
| HQ687756.1 | - | 1 - 2 | 427 | 119 | ||
| J05228.1 | - | 1 - 2 | 302 | 119 | ||
More Information
Set Membership: |
|

IGFBP3
Genomic Map