Assay Details
| Gene Symbol: SLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044566.2 | NP_001038031.1 | 2 - 3 | 283 | 80 | |
| GenBank mRNA | AK347267.1 | - | 1 - 2 | 178 | 80 | |
| DQ456976.1 | - | 1 - 2 | 95 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001044566.2 transcripts.
Set Membership: |
|

SLN
Genomic Map