Assay Details
| Gene Symbol: TFB1M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128475.1 | NP_001121947.1 | 6 - 7 | 794 | 76 | |
| XM_021083526.1 | XP_020939185.1 | 745 | 76 | |||
| GenBank mRNA | AK236183.1 | - | 6 - 7 | 854 | 76 | |
| EU617322.1 | - | 6 - 7 | 794 | 76 | ||
More Information
Set Membership: |
|

TFB1M
Genomic Map