Assay Details
| Gene Symbol: CRABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128492.1 | NP_001121964.1 | 1 - 2 | 73 | 96 | |
| GenBank mRNA | AY610183.1 | - | 1 - 2 | 178 | 96 | |
| EF397416.1 | - | 1 - 2 | 73 | 96 | ||
More Information
Set Membership: |
|

CRABP1
Genomic Map