Assay Details
| Gene Symbol: UQCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129978.2 | NP_001123450.1 | 1 - 2 | 174 | 91 | |
| GenBank mRNA | AK231574.1 | - | 1 - 2 | 164 | 91 | |
| AK344853.1 | - | 1 - 2 | 168 | 91 | ||
| DQ372078.1 | - | 1 - 2 | 175 | 91 | ||
More Information
Set Membership: |
|

UQCR10
Genomic Map