Assay Details
| Gene Symbol: SLC7A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130973.1 | NP_001124445.1 | 11 - 12 | 1851 | 66 | |
| GenBank mRNA | EU780707.1 | - | 11 - 12 | 1851 | 66 | |
More Information
Set Membership: |
|

SLC7A3
Genomic Map