Assay Details
| Gene Symbol: PRPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135965.1 | NP_001129437.1 | 6 - 7 | 802 | 91 | |
| XM_005655388.3 | XP_005655445.1 | 991 | 91 | |||
| GenBank mRNA | AK346062.1 | - | 6 - 7 | 797 | 91 | |
| AK392258.1 | - | 6 - 7 | 819 | 91 | ||
| AK393632.1 | - | 6 - 7 | 813 | 91 | ||
| FJ167679.1 | - | 6 - 7 | 802 | 91 | ||
More Information
Set Membership: |
|

PRPF3
Genomic Map