Assay Details
| Gene Symbol: NCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137630.1 | NP_001131102.1 | 2 - 3 | 937 | 73 | |
| XM_005669868.2 | XP_005669925.1 | 1257 | 73 | |||
| XM_021068315.1 | XP_020923974.1 | 1114 | 73 | |||
| XM_021068316.1 | XP_020923975.1 | 1442 | 73 | |||
| GenBank mRNA | AK230672.1 | - | 2 - 3 | 810 | 73 | |
| AK233065.1 | - | 3 - 4 | 1037 | 73 | ||
| AK392504.1 | - | 3 - 4 | 1057 | 73 | ||
| AY610196.1 | - | 948 | 73 | |||
| FJ200452.1 | - | 2 - 3 | 937 | 73 | ||
More Information
Set Membership: |
|

NCK1
Genomic Map