Assay Details
| Gene Symbol: DBF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137637.1 | NP_001131109.1 | 6 - 7 | 813 | 68 | |
| XM_021101967.1 | XP_020957626.1 | 936 | 68 | |||
| XM_021101968.1 | XP_020957627.1 | 878 | 68 | |||
| XM_021101969.1 | XP_020957628.1 | 937 | 68 | |||
| XM_021101970.1 | XP_020957629.1 | 878 | 68 | |||
| XM_021101971.1 | XP_020957630.1 | 939 | 68 | |||
| GenBank mRNA | FJ211192.1 | - | 6 - 7 | 813 | 68 | |
More Information
Set Membership: |
|

DBF4
Genomic Map