Assay Details
| Gene Symbol: TNFRSF12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142839.2 | NP_001136311.1 | 3 - 4 | 355 | 92 | |
| GenBank mRNA | AY609823.1 | - | 3 - 4 | 349 | 92 | |
| EU417836.1 | - | 3 - 4 | 355 | 92 | ||
More Information
Set Membership: |
|

TNFRSF12A
Genomic Map