Assay Details
| Gene Symbol: LGALS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142842.1 | NP_001136314.1 | 1 - 2 | 33 | 69 | |
| GenBank mRNA | AK231447.1 | - | 1 - 2 | 65 | 69 | |
| FJ424704.1 | - | 1 - 2 | 33 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001142842.1 transcripts.
Set Membership: |
|

LGALS2
Genomic Map