Assay Details
| Gene Symbol: PKMYT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159310.1 | NP_001152782.1 | 2 - 3 | 415 | 60 | |
| XM_021085610.1 | XP_020941269.1 | 769 | 60 | |||
| XM_021085611.1 | XP_020941270.1 | 687 | 60 | |||
| XM_021085612.1 | XP_020941271.1 | 572 | 60 | |||
| XM_021085613.1 | XP_020941272.1 | 815 | 60 | |||
| XM_021085614.1 | XP_020941273.1 | 775 | 60 | |||
| XM_021085615.1 | XP_020941274.1 | 960 | 60 | |||
| XM_021085616.1 | XP_020941275.1 | 693 | 60 | |||
| GenBank mRNA | AB495207.1 | - | 2 - 3 | 415 | 60 | |
More Information
Set Membership: |
|

PKMYT1
Genomic Map