Assay Details
| Gene Symbol: MOGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167651.1 | NP_001161123.1 | 5 - 6 | 852 | 88 | |
| XM_013979136.2 | XP_013834590.1 | 2487 | 88 | |||
| GenBank mRNA | AB529861.1 | - | 5 - 6 | 852 | 88 | |
More Information
Set Membership: |
|

MOGAT2
Genomic Map