Assay Details
| Gene Symbol: CSRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172368.1 | NP_001165839.1 | 4 - 5 | 517 | 64 | |
| GenBank mRNA | AY610264.1 | - | 3 - 4 | 451 | 64 | |
| FJ494917.1 | - | 4 - 5 | 517 | 64 | ||
More Information
Set Membership: |
|

CSRP3
Genomic Map