Assay Details
| Gene Symbol: SPATA2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177918.1 | NP_001171389.1 | 1 - 2 | 302 | 68 | |
| GenBank mRNA | GU475010.1 | - | 1 - 2 | 302 | 68 | |
More Information
Set Membership: |
|

SPATA2L
Genomic Map