Assay Details
| Gene Symbol: TRIM63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184756.1 | NP_001171685.1 | 1 - 2 | 257 | 56 | |
| GenBank mRNA | FJ905227.1 | - | 1 - 2 | 257 | 56 | |
More Information
Set Membership: |
|

TRIM63
Genomic Map