Assay Details
| Gene Symbol: MARCHF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244915.1 | NP_001231844.1 | 2 - 3 | 318 | 77 | |
| XM_005654752.3 | XP_005654809.1 | 370 | 77 | |||
| GenBank mRNA | AK236835.1 | - | 2 - 3 | 318 | 77 | |
| AK396059.1 | - | 2 - 3 | 324 | 77 | ||
| AY609952.1 | - | 2 - 3 | 318 | 77 | ||
More Information
Set Membership: |
|

MARCHF2
Genomic Map