Assay Details
| Gene Symbol: MYCBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244917.1 | NP_001231846.1 | 4 - 5 | 332 | 103 | |
| GenBank mRNA | AK236342.1 | - | 4 - 5 | 332 | 103 | |
| AY610032.1 | - | 4 - 5 | 308 | 103 | ||
More Information
Set Membership: |
|

MYCBP
Genomic Map