Assay Details
| Gene Symbol: MTRF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244934.1 | NP_001231863.1 | 4 - 5 | 708 | 89 | |
| XM_013992473.2 | XP_013847927.2 | 3613 | 89 | |||
| XM_013992474.2 | XP_013847928.1 | 585 | 89 | |||
| XM_021072417.1 | XP_020928076.1 | 602 | 89 | |||
| GenBank mRNA | AK231749.1 | - | 4 - 5 | 427 | 89 | |
| AY609949.1 | - | 4 - 5 | 708 | 89 | ||
More Information
Set Membership: |
|

MTRF1L
Genomic Map