Assay Details
| Gene Symbol: MRPL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244967.1 | NP_001231896.1 | 1 - 2 | 173 | 104 | |
| GenBank mRNA | AK237180.1 | - | 1 - 2 | 173 | 104 | |
| AY609729.1 | - | 1 - 2 | 147 | 104 | ||
More Information
Set Membership: |
|

MRPL16
Genomic Map