Assay Details
| Gene Symbol: KDELR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244970.1 | NP_001231899.1 | 1 - 2 | 263 | 70 | |
| GenBank mRNA | AK236601.1 | - | 1 - 2 | 263 | 70 | |
| AK397568.1 | - | 1 - 2 | 177 | 70 | ||
More Information
Set Membership: |
|

KDELR3
Genomic Map