Assay Details
| Gene Symbol: KCTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244971.2 | NP_001231900.2 | 4 - 5 | 2459 | 61 | |
| XM_005653406.3 | XP_005653463.1 | 1566 | 61 | |||
| XM_005653407.3 | XP_005653464.1 | 662 | 61 | |||
| XM_021093325.1 | XP_020948984.1 | 2405 | 61 | |||
| GenBank mRNA | AK395469.1 | - | 4 - 5 | 1058 | 61 | |
| AY610046.1 | - | 4 - 5 | 768 | 61 | ||
More Information
Set Membership: |
|

KCTD1
Genomic Map