Assay Details
| Gene Symbol: VBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244980.1 | NP_001231909.1 | 3 - 4 | 295 | 137 | |
| XM_005674019.3 | XP_005674076.1 | 398 | 137 | |||
| GenBank mRNA | AK230843.1 | - | 3 - 4 | 295 | 137 | |
| AK392033.1 | - | 3 - 4 | 308 | 137 | ||
| AK399546.1 | - | 3 - 4 | 318 | 137 | ||
More Information
Set Membership: |
|

VBP1
Genomic Map