Assay Details
| Gene Symbol: SEPTIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244989.1 | NP_001231918.1 | 5 - 6 | 473 | 78 | |
| XM_021085671.1 | XP_020941330.1 | 415 | 78 | |||
| GenBank mRNA | AK237018.1 | - | 5 - 6 | 473 | 78 | |
| AK238762.1 | - | 4 - 5 | 359 | 78 | ||
| AY609759.1 | - | 3 - 4 | 378 | 78 | ||
| KU358533.1 | - | 4 - 5 | 375 | 78 | ||
More Information
Set Membership: |
|

SEPTIN1
Genomic Map