Assay Details
| Gene Symbol: SEPTIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244989.1 | NP_001231918.1 | 7 - 8 | 717 | 74 | |
| XM_021085671.1 | XP_020941330.1 | 659 | 74 | |||
| GenBank mRNA | AK237018.1 | - | 7 - 8 | 717 | 74 | |
| AK238762.1 | - | 6 - 7 | 603 | 74 | ||
| AY609759.1 | - | 5 - 6 | 622 | 74 | ||
| KU358533.1 | - | 6 - 7 | 619 | 74 | ||
More Information
Set Membership: |
|

SEPTIN1
Genomic Map