Assay Details
| Gene Symbol: SEPTIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244989.1 | NP_001231918.1 | 8 - 9 | 824 | 83 | |
| XM_021085671.1 | XP_020941330.1 | 766 | 83 | |||
| GenBank mRNA | AK237018.1 | - | 8 - 9 | 824 | 83 | |
| AK238762.1 | - | 7 - 8 | 710 | 83 | ||
| KU358533.1 | - | 7 - 8 | 726 | 83 | ||
More Information
Set Membership: |
|

SEPTIN1
Genomic Map