Assay Details
| Gene Symbol: SEPTIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244989.1 | NP_001231918.1 | 9 - 10 | 915 | 109 | |
| XM_021085671.1 | XP_020941330.1 | 857 | 109 | |||
| GenBank mRNA | AK237018.1 | - | 9 - 10 | 915 | 109 | |
| AY609759.1 | - | 7 - 8 | 820 | 109 | ||
| KU358533.1 | - | 8 - 9 | 817 | 109 | ||
More Information
Set Membership: |
|

SEPTIN1
Genomic Map