Assay Details
Gene Symbol: CLUAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245000.1 | NP_001231929.1 | 10 - 11 | 1106 | 56 | |
XM_021085672.1 | XP_020941331.1 | 1464 | 56 | |||
XM_021085673.1 | XP_020941332.1 | 1343 | 56 | |||
XM_021085674.1 | XP_020941333.1 | 1339 | 56 | |||
GenBank mRNA | AK346084.1 | - | 10 - 11 | 1106 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
