Assay Details
| Gene Symbol: PLPPR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001245001.1 | NP_001231930.1 | 1 - 2 | 334 | 84 | |
| XM_013997347.2 | XP_013852801.1 | 580 | 84 | |||
| XM_021088656.1 | XP_020944315.1 | 1390 | 84 | |||
| XM_021088657.1 | XP_020944316.1 | 1389 | 84 | |||
| GenBank mRNA | AK343398.1 | - | 1 - 2 | 334 | 84 | |
More Information
Set Membership: |
|

PLPPR5
Genomic Map