Assay Details
| Gene Symbol: ZNF232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001245009.1 | NP_001231938.1 | 3 - 4 | 1051 | 81 | |
| XM_005669149.3 | XP_005669206.1 | 1251 | 81 | |||
| XM_005669150.3 | XP_005669207.1 | 1143 | 81 | |||
| XM_005669151.3 | XP_005669208.1 | 1111 | 81 | |||
| XM_005669152.3 | XP_005669209.1 | 684 | 81 | |||
| XM_021066110.1 | XP_020921769.1 | 1331 | 81 | |||
| XM_021066111.1 | XP_020921770.1 | 1223 | 81 | |||
| XM_021066113.1 | XP_020921772.1 | 1159 | 81 | |||
| XM_021066114.1 | XP_020921773.1 | 1022 | 81 | |||
| GenBank mRNA | AK349548.1 | - | 3 - 4 | 1051 | 81 | |
More Information
Set Membership: |
|

ZNF232
Genomic Map