Assay Details
| Gene Symbol: RNF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001245011.1 | NP_001231940.1 | 1 - 2 | 318 | 71 | |
| GenBank mRNA | AK240441.1 | - | 1 - 2 | 318 | 71 | |
| AK395453.1 | - | 1 - 2 | 321 | 71 | ||
| AY609394.1 | - | 1 - 2 | 297 | 71 | ||
More Information
Set Membership: |
|

RNF7
Genomic Map