Assay Details
| Gene Symbol: RNF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001245011.1 | NP_001231940.1 | 2 - 3 | 371 | 79 | |
| XM_005669903.3 | XP_005669960.1 | 219 | 79 | |||
| GenBank mRNA | AK240441.1 | - | 2 - 3 | 371 | 79 | |
| AK395453.1 | - | 2 - 3 | 374 | 79 | ||
| AY609394.1 | - | 2 - 3 | 350 | 79 | ||
More Information
Set Membership: |
|

RNF7
Genomic Map