Assay Details
| Gene Symbol: IMMP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001245014.1 | NP_001231943.1 | 5 - 6 | 572 | 111 | |
| XM_005661044.3 | XP_005661101.1 | 607 | 111 | |||
| XM_005661046.3 | XP_005661103.1 | 545 | 111 | |||
| XM_005661047.3 | XP_005661104.1 | 466 | 111 | |||
| XM_005661048.3 | XP_005661105.1 | 466 | 111 | |||
| XM_013994453.2 | XP_013849907.1 | 474 | 111 | |||
| XM_021078971.1 | XP_020934630.1 | 372 | 111 | |||
| XM_021078985.1 | XP_020934644.1 | 411 | 111 | |||
| GenBank mRNA | AK347172.1 | - | 5 - 6 | 572 | 111 | |
More Information
Set Membership: |
|

IMMP1L
Genomic Map