Assay Details
| Gene Symbol: PQBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246668.1 | NP_001233597.1 | 2 - 3 | 203 | 63 | |
| XM_005673594.3 | XP_005673651.1 | 347 | 63 | |||
| XM_013986114.1 | XP_013841568.1 | 613 | 63 | |||
| GenBank mRNA | AK231983.1 | - | 3 - 4 | 339 | 63 | |
| AK238508.1 | - | 3 - 4 | 618 | 63 | ||
| AK351316.1 | - | 3 - 4 | 616 | 63 | ||
| AK352148.1 | - | 3 - 4 | 339 | 63 | ||
| AK397584.1 | - | 3 - 4 | 342 | 63 | ||
| AY609811.1 | - | 247 | 63 | |||
| JF750401.1 | - | 2 - 3 | 203 | 63 | ||
More Information
Set Membership: |
|

PQBP1
Genomic Map