Assay Details
| Gene Symbol: LOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206403.1 | NP_001193332.1 | 2 - 3 | 766 | 84 | |
| GenBank mRNA | AK395814.1 | - | 2 - 3 | 766 | 84 | |
| CK452593.1 | - | 5 - 6 | 816 | 84 | ||
More Information
Set Membership: |
|

LOX
Genomic Map