Assay Details
| Gene Symbol: GALNT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206447.1 | NP_001193376.1 | 3 - 4 | 1433 | 82 | |
| XM_013981389.2 | XP_013836843.2 | 1698 | 82 | |||
| XM_021068276.1 | XP_020923935.1 | 1669 | 82 | |||
| GenBank mRNA | AK351123.1 | - | 3 - 4 | 1431 | 82 | |
| AK389431.1 | - | 3 - 4 | 860 | 82 | ||
More Information
Set Membership: |
|

GALNT15
Genomic Map