Assay Details
| Gene Symbol: LIPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243029.1 | NP_001229958.1 | 4 - 5 | 897 | 58 | |
| XM_013992843.2 | XP_013848297.1 | 891 | 58 | |||
| XM_021094488.1 | XP_020950147.1 | 858 | 58 | |||
| GenBank mRNA | AK343278.1 | - | 4 - 5 | 897 | 58 | |
More Information
Set Membership: |
|

LIPG
Genomic Map