Assay Details
| Gene Symbol: MAP1S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243185.1 | NP_001230114.1 | 5 - 6 | 2835 | 61 | |
| XM_021079065.1 | XP_020934724.1 | 3419 | 61 | |||
| GenBank mRNA | AK235554.1 | - | 5 - 6 | 2813 | 61 | |
| AK391745.1 | - | 5 - 6 | 2825 | 61 | ||
More Information
Set Membership: |
|

MAP1S
Genomic Map