Assay Details
| Gene Symbol: SDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243190.1 | NP_001230119.1 | 1 - 2 | 410 | 96 | |
| GenBank mRNA | AK345304.1 | - | 1 - 2 | 410 | 96 | |
| AK394158.1 | - | 1 - 2 | 406 | 96 | ||
More Information
Set Membership: |
|

SDC1
Genomic Map