Assay Details
| Gene Symbol: NIBAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243219.1 | NP_001230148.1 | 12 - 13 | 1724 | 103 | |
| GenBank mRNA | AK236453.1 | - | 1 - 2 | 108 | 103 | |
| AK346027.1 | - | 12 - 13 | 1724 | 103 | ||
| AK347893.1 | - | 12 - 13 | 1688 | 103 | ||
| AK399056.1 | - | 1 - 2 | 47 | 103 | ||
More Information
Set Membership: |
|

NIBAN1
Genomic Map