Assay Details
| Gene Symbol: NIBAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243219.1 | NP_001230148.1 | 13 - 14 | 1840 | 117 | |
| GenBank mRNA | AK236453.1 | - | 2 - 3 | 224 | 117 | |
| AK346027.1 | - | 13 - 14 | 1840 | 117 | ||
| AK347893.1 | - | 13 - 14 | 1804 | 117 | ||
| AK399056.1 | - | 2 - 3 | 163 | 117 | ||
More Information
Set Membership: |
|

NIBAN1
Genomic Map