Assay Details
| Gene Symbol: PSPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243221.1 | NP_001230150.1 | 6 - 7 | 902 | 67 | |
| XM_005662001.2 | XP_005662058.1 | 848 | 67 | |||
| XM_021085632.1 | XP_020941291.1 | 666 | 67 | |||
| GenBank mRNA | AB530144.1 | - | 4 - 5 | 587 | 67 | |
| AJ963844.1 | - | 1 - 2 | 523 | 67 | ||
| AK233338.1 | - | 6 - 7 | 902 | 67 | ||
| AK348175.1 | - | 6 - 7 | 809 | 67 | ||
| AY610038.1 | - | 6 - 7 | 891 | 67 | ||
| DN106508.1 | - | 1 - 2 | 497 | 67 | ||
More Information
Set Membership: |
|

PSPH
Genomic Map