Assay Details
Gene Symbol: FHL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243314.1 | NP_001230243.1 | 2 - 3 | 252 | 64 | |
XM_021084440.1 | XP_020940099.1 | 631 | 64 | |||
XM_021084477.1 | XP_020940136.1 | 886 | 64 | |||
GenBank mRNA | AK345676.1 | - | 2 - 3 | 334 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
