Assay Details
| Gene Symbol: SPSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243315.1 | NP_001230244.1 | 1 - 2 | 173 | 75 | |
| GenBank mRNA | AK349141.1 | - | 1 - 2 | 173 | 75 | |
| AK400606.1 | - | 1 - 2 | 174 | 75 | ||
| FS690733.1 | - | 1 - 2 | 174 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243315.1 transcripts.
Set Membership: |
|

SPSB2
Genomic Map