Assay Details
| Gene Symbol: HEY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243329.1 | NP_001230258.1 | 3 - 4 | 393 | 98 | |
| XM_013992576.2 | XP_013848030.1 | 292 | 98 | |||
| XM_021086293.1 | XP_020941952.1 | 1092 | 98 | |||
| GenBank mRNA | BP149699.1 | - | 3 - 4 | 394 | 98 | |
More Information
Set Membership: |
|

HEY2
Genomic Map